Learn More
Shockwatch Div Mri WarmMark Duo 10C/50F-34C/93F
SDP
Catalog No. NC2598312
Encompass_Preferred
WarmMark Duo time-temperature indicator monitors two temperatures during shipping and storage, indicating if goods experience unacceptable conditions. These reliable temperature indicators provide visual evidence of exposure to unacceptably high temperature levels, allowing you to determine if product quality has been compromised. Monitor time above the device threshold and indicate if temperature exceeded for the specified time. Indicators are field-armable by pulling a tab. Temperature threshold: 10C/50F and 34C/. Accuracy +/-1C. Above temperature exposure time: 14 days (10C/50F) and within 30 minutes 34C/93F. Shelf life 2 years. Single-use.
Catalog No. NC2598312 Supplier Shockwatch Div Mri Supplier No. WD 1034
WarmMark Duo time-temperature indicator monitors two temperatures during shipping and storage, indicating if goods experience unacceptable conditions. These reliable temperature indicators provide visual evidence of exposure to unacceptably high temperature levels, allowing you to determine if product quality has been compromised. Monitor time above the device threshold and indicate if temperature exceeded for the specified time. Indicators are field-armable by pulling a tab. Temperature threshold: 10C/50F and 34C/. Accuracy +/-1C. Above temperature exposure time: 14 days (10C/50F) and within 30 minutes 34C/93F. Shelf life 2 years. Single-use.