Learn More
GA International Warning Labels - Biohazard 1.125" x 1.125" / 28.6mm x 28.6mm Rectangle Black on Yellow From -80°C to +100°C (-112°F to +212°F)
Catalog No. 5000571326
Encompass_Preferred
These pre-printed warning labels with a bold, universally recognized trefoil biohazard symbol on a bright yellow background are suitable for labeling plastic bags, tubes, cardboard, and other boxes containing biohazard materials such as pathological waste and used hypodermic needles. These durable labels can resist abrasion and exposure to harsh chemicals and solvents as well as a wide range of temperatures, from -80°C to +100°C. Our water and tearproof warning labels are designed to help biological laboratories communicate potential hazards and minimize the risk of exposure to hazardous biological materials.
Catalog No. 50-005-71326 Supplier GA International Supplier No. WLA0071P
These pre-printed warning labels with a bold, universally recognized trefoil biohazard symbol on a bright yellow background are suitable for labeling plastic bags, tubes, cardboard, and other boxes containing biohazard materials such as pathological waste and used hypodermic needles. These durable labels can resist abrasion and exposure to harsh chemicals and solvents as well as a wide range of temperatures, from -80°C to +100°C. Our water and tearproof warning labels are designed to help biological laboratories communicate potential hazards and minimize the risk of exposure to hazardous biological materials.