Learn More
GA International Warning Labels for Deep-Freeze Application - "Biohazard Symbol" 0.5" / 12.7mm Circle Black on Red From -80°C to +100°C (-112°F to +212°F)
Catalog No. 5000571702
Encompass_Preferred
Biohazard dot labels are laminated plastic tube labels for identifying vials, tubes, and other containers with a biohazard symbol. These round biohazard labels are ideal for prolonged storage inside freezers (-80°C), and dry ice, as well as for transportation of hazardous biological samples and other hazardous materials in dry ice containers, and portable freezers. These warning labels can withstand multiple freeze-thaw cycles, boiling water, dry ice, and other cryogenic and harsh lab environments. The radioactive symbol and the colored background are protected against water, alcohols, xylene, toluene, acetone, DMSO, MEK, industrial thinners, oil, grease, acids, bases, brake fluid, detergents, and other chemicals and solvents, as well as mechanical friction, rubbing, under the hood use and physical damage. Wear and tear proof.
Catalog No. 50-005-71702 Supplier GA International Supplier No. WL0121P
Biohazard dot labels are laminated plastic tube labels for identifying vials, tubes, and other containers with a biohazard symbol. These round biohazard labels are ideal for prolonged storage inside freezers (-80°C), and dry ice, as well as for transportation of hazardous biological samples and other hazardous materials in dry ice containers, and portable freezers. These warning labels can withstand multiple freeze-thaw cycles, boiling water, dry ice, and other cryogenic and harsh lab environments. The radioactive symbol and the colored background are protected against water, alcohols, xylene, toluene, acetone, DMSO, MEK, industrial thinners, oil, grease, acids, bases, brake fluid, detergents, and other chemicals and solvents, as well as mechanical friction, rubbing, under the hood use and physical damage. Wear and tear proof.