Learn More
GA International Warning Labels for Deep-Freeze Application - "Radioactive Symbol" 0.5" / 12.7mm Circle Black on Yellow From -80°C to +100°C (-112°F to +212°F)
Catalog No. 5000571432
Encompass_Preferred
Thermoplastic dot labels, with the internationally recognized trefoil black symbol on a yellow background, for the identification of vials, tubes, as well as other containers and surfaces that contain or may have been in contact with radiation-emitting materials. These round radioactivity warning labels are suitable for prolonged storage in deep-freeze conditions, as well as transportation of radioactive material, such as radioisotopes, in dry ice containers and portable freezers. The waterproof, tearproof, warning labels will resist exposure to harsh chemicals and solvents, as well as mechanical friction, rubbing, and physical damage.
Catalog No. 50-005-71432 Supplier GA International Supplier No. WL0131P
Thermoplastic dot labels, with the internationally recognized trefoil black symbol on a yellow background, for the identification of vials, tubes, as well as other containers and surfaces that contain or may have been in contact with radiation-emitting materials. These round radioactivity warning labels are suitable for prolonged storage in deep-freeze conditions, as well as transportation of radioactive material, such as radioisotopes, in dry ice containers and portable freezers. The waterproof, tearproof, warning labels will resist exposure to harsh chemicals and solvents, as well as mechanical friction, rubbing, and physical damage.