Learn More
GA International Warning Labels for Deep-Freeze Application - "Stop Sign" 1.5" x 1.5" / 38.1mm x 38.1mm Rectangle White on Red From -80°C to +100°C (-112°F to +212°F)
Catalog No. 5000571167
Encompass_Preferred
Pre-printed octagonal red stop sign warning labels. These warning labels are waterproof and tearproof, able to withstand low temperature environments, in laboratory freezers (-80°C), and transportation on dry ice. The labels are able to resist abrasion and exposure to harsh chemicals and solvents, ideal for effectively alerting users of safety procedures in a variety of environments. They can also be applied on refrigerated and frozen (-80°C/-112°F) plastic and cardboard surfaces.
Catalog No. 50-005-71167 Supplier GA International Supplier No. WL0111P
Pre-printed octagonal red stop sign warning labels. These warning labels are waterproof and tearproof, able to withstand low temperature environments, in laboratory freezers (-80°C), and transportation on dry ice. The labels are able to resist abrasion and exposure to harsh chemicals and solvents, ideal for effectively alerting users of safety procedures in a variety of environments. They can also be applied on refrigerated and frozen (-80°C/-112°F) plastic and cardboard surfaces.