Learn More
GA International Warning Labels for Deep-Freeze Applications - "Radioactive Symbol" 0.5" / 12.7mm Circle Red on Yellow From -80°C to +110°C (-112°F to +230°F)
Catalog No. 5000571704
Encompass_Preferred
Thermoplastic dot labels, with the red internationally recognized trefoil symbol on a yellow background, for the identification of vials, tubes as well as other containers and surfaces that contain or may have been in contact with radiation-emitting materials. These round radioactivity warning labels are suitable for prolonged storage in deep-freeze conditions, as well as transportation of radioactive material, such as radioisotopes, in dry ice containers, and portable freezers. The waterproof, tearproof, warning labels will resist exposure to harsh chemical and solvents, as well as mechanical friction, rubbing, and physical damage.
Catalog No. 50-005-71704 Supplier GA International Supplier No. WLA0141P
Thermoplastic dot labels, with the red internationally recognized trefoil symbol on a yellow background, for the identification of vials, tubes as well as other containers and surfaces that contain or may have been in contact with radiation-emitting materials. These round radioactivity warning labels are suitable for prolonged storage in deep-freeze conditions, as well as transportation of radioactive material, such as radioisotopes, in dry ice containers, and portable freezers. The waterproof, tearproof, warning labels will resist exposure to harsh chemical and solvents, as well as mechanical friction, rubbing, and physical damage.