Learn More
GA International Warning Labels for General Laboratory Use - "Radioactive" 2.72" x 1" / 69.1mm x 25.4mm Rectangle Red text on Yellow Ambient temperature
Catalog No. 5000570976
Encompass_Preferred
Standard warning labels to identify radioactive samples and materials. Paper labels leave no residue behind after removal and are for use at ambient temperatures.
Catalog No. 50-005-70976 Supplier GA International Supplier No. L0041R
Standard warning labels to identify radioactive samples and materials. Paper labels leave no residue behind after removal and are for use at ambient temperatures.