Learn More
GA International Warning Labels - "Non Hazardous" 1.75" x 0.75" / 44.5mm x 19.1mm Rectangle Black on Mint From -80°C to +100°C (-112°F to +212°F)
Catalog No. 5000571873
Encompass_Preferred
Deep-freeze pre-printed thermoplastic warning labels identifying containers and samples as non-hazardous. These freezer labels are waterproof, tearproof, can withstand multiple freeze-thaw cycles, transportation on dry ice, long-term storage in low temperature freezers, and other deep-freeze lab environments. They can resist exposure to various harsh chemicals and solvents, as well as abrasion.
Catalog No. 50-005-71873 Supplier GA International Supplier No. WL0020.25P
Deep-freeze pre-printed thermoplastic warning labels identifying containers and samples as non-hazardous. These freezer labels are waterproof, tearproof, can withstand multiple freeze-thaw cycles, transportation on dry ice, long-term storage in low temperature freezers, and other deep-freeze lab environments. They can resist exposure to various harsh chemicals and solvents, as well as abrasion.