Learn More
GA International Warning Labels - "Toxic Warning Symbol" 1.75" x 0.75" / 44.5mm x 19.1mm Rectangle Black on Red From -80°C to +100°C (-112°F to +212°F)
Catalog No. 5000571966
Encompass_Preferred
Deep-freeze pre-printed thermoplastic warning labels identifying the presence of toxic substances. These freezer-safe labels have a special construct that protects the printout from abrasion and exposure to various harsh chemicals and solvents. Perfect for use in laboratory freezers and refrigerators in research facilities, hospitals, clinics, as well as environmental testing companies. Water-resistant, wear and tear proof, the labels can be used to identify plastic and glass bottles, boxes, tubes, and other containers.
Catalog No. 50-005-71966 Supplier GA International Supplier No. WL0010.25P
Deep-freeze pre-printed thermoplastic warning labels identifying the presence of toxic substances. These freezer-safe labels have a special construct that protects the printout from abrasion and exposure to various harsh chemicals and solvents. Perfect for use in laboratory freezers and refrigerators in research facilities, hospitals, clinics, as well as environmental testing companies. Water-resistant, wear and tear proof, the labels can be used to identify plastic and glass bottles, boxes, tubes, and other containers.