Learn More
Grainger Edwards Signaling Warning Light, LED, 120V AC, Green, 1 ea
Catalog No. 502790783
Encompass_Preferred
This heavy-duty LED beacon is designed for reliable visual signaling in adverse, hazardous, or marine environments. The unit features multi-mode operation, providing either a steady-on light or a flashing signal at 65 fpm, which is field-selectable via dipswitch. Constructed with a shatter-resistant polycarbonate lens and a glass-reinforced thermoplastic polyester base, it is built to withstand demanding industrial conditions while remaining immune to shock and vibration.
  • Provides reliable visual alarms in hazardous locations including Class I, Div 2, Groups A, B, C, and D; Class II, Div 2, Groups F and G; and Class III
  • Features a NEMA 4X, Type 4X rated enclosure for protection against harsh elements
  • Offers an operational life of 148,000 hours
  • Supports flexible mounting options including 3/4 in. NPT pipe, junction box, or wall mount configurations
  • Operates at 120 V AC with a current draw of
Catalog No. 50-279-0783 Supplier Grainger Supplier No. 16G643
This heavy-duty LED beacon is designed for reliable visual signaling in adverse, hazardous, or marine environments. The unit features multi-mode operation, providing either a steady-on light or a flashing signal at 65 fpm, which is field-selectable via dipswitch. Constructed with a shatter-resistant polycarbonate lens and a glass-reinforced thermoplastic polyester base, it is built to withstand demanding industrial conditions while remaining immune to shock and vibration.
  • Provides reliable visual alarms in hazardous locations including Class I, Div 2, Groups A, B, C, and D; Class II, Div 2, Groups F and G; and Class III
  • Features a NEMA 4X, Type 4X rated enclosure for protection against harsh elements
  • Offers an operational life of 148,000 hours
  • Supports flexible mounting options including 3/4 in. NPT pipe, junction box, or wall mount configurations
  • Operates at 120 V AC with a current draw of