Learn More
Grainger Edwards Signaling Multi-Mode LED Warning Light, Green, 120V AC
Catalog No. 502788765
Encompass_Preferred
This heavy-duty, multi-mode LED signaling beacon is designed for indoor and outdoor warning applications. It features a green lens and a durable gray Rynite® (PET) base, providing reliable performance in a variety of industrial environments. The unit operates on 120 V AC and offers versatile functionality, including user-selectable steady-on or flashing modes.
  • NEMA 4X rated enclosure for protection against harsh environments
  • Features long-lasting LED technology with a median lifespan of 148,000 hours
  • Mounting flexibility via 1/2 in. internal or 3/4 in. external conduit threading
  • Wide operating temperature range from -35°C to +66°C (-31°F to +150°F)
  • UL listed for marine applications and hazardous locations including Class I, Div 2
  • Operational at 50/60 Hz with 0.108 A current draw
Catalog No. 50-278-8765 Supplier Grainger Supplier No. 16G629
This heavy-duty, multi-mode LED signaling beacon is designed for indoor and outdoor warning applications. It features a green lens and a durable gray Rynite® (PET) base, providing reliable performance in a variety of industrial environments. The unit operates on 120 V AC and offers versatile functionality, including user-selectable steady-on or flashing modes.
  • NEMA 4X rated enclosure for protection against harsh environments
  • Features long-lasting LED technology with a median lifespan of 148,000 hours
  • Mounting flexibility via 1/2 in. internal or 3/4 in. external conduit threading
  • Wide operating temperature range from -35°C to +66°C (-31°F to +150°F)
  • UL listed for marine applications and hazardous locations including Class I, Div 2
  • Operational at 50/60 Hz with 0.108 A current draw