Learn More
Grainger Edwards Signaling Beacon Light, LED, Red, 120 VAC, 1 ea
Catalog No. 502789572
Encompass_Preferred
The 125 Class XBR LED Beacon is a dual-mode signaling appliance designed for high-performance visual notification. Rated for NEMA 4X applications, this beacon features an XTRA-BRIGHT LED light source housed in a durable 33% glass-filled nylon base with a shatter-resistant polycarbonate lens. It operates at 120 VAC and offers selectable modes, allowing users to switch from steady-on to lightburst mode to maximize visibility in critical environments. The unit is designed for versatile installation, supporting both surface and conduit mounting.
  • Dual mode operation with steady-on and lightburst settings
  • NEMA 4X enclosure for corrosion, heat, and chemical resistance
  • Long operational life rated at 148,000 hr
  • Supports 1/2 in. internal and 3/4 in. external conduit hub mounting
  • UL and cUL listed for safety and reliability
  • Operating temperature range from -35°C to 66°C (-31°F to 15
Catalog No. 50-278-9572 Supplier Grainger Supplier No. 16G634
The 125 Class XBR LED Beacon is a dual-mode signaling appliance designed for high-performance visual notification. Rated for NEMA 4X applications, this beacon features an XTRA-BRIGHT LED light source housed in a durable 33% glass-filled nylon base with a shatter-resistant polycarbonate lens. It operates at 120 VAC and offers selectable modes, allowing users to switch from steady-on to lightburst mode to maximize visibility in critical environments. The unit is designed for versatile installation, supporting both surface and conduit mounting.
  • Dual mode operation with steady-on and lightburst settings
  • NEMA 4X enclosure for corrosion, heat, and chemical resistance
  • Long operational life rated at 148,000 hr
  • Supports 1/2 in. internal and 3/4 in. external conduit hub mounting
  • UL and cUL listed for safety and reliability
  • Operating temperature range from -35°C to 66°C (-31°F to 15