Learn More
Grainger Edwards Signaling Warning Light, Red LED, 120V AC, NEMA 4X, 1 ea
Catalog No. 502789886
Encompass_Preferred
This Xtra-Brite series warning light provides a reliable visual signaling solution in a compact, durable design. It is engineered for both steady-on and flashing operation, making it suitable for a variety of industrial alerting applications. The unit features an energy-efficient LED light source housed within a corrosion-resistant, NEMA 4X rated enclosure, ensuring protection in harsh environments.
  • Features a long-lasting LED light source with a 148,000 hr median life
  • Configurable for steady burning or flashing operation with a flash rate of 65 fpm
  • Constructed with a shatter-resistant polycarbonate lens and a 33% glass-filled nylon base
  • Rated NEMA 4X for corrosion resistance
  • Designed for conduit or panel mounting with 1/2 in. internal and 3/4 in. external hub threading
  • Immune to shock and vibration
  • UL and cUL listed
Catalog No. 50-278-9886 Supplier Grainger Supplier No. 16G613
This Xtra-Brite series warning light provides a reliable visual signaling solution in a compact, durable design. It is engineered for both steady-on and flashing operation, making it suitable for a variety of industrial alerting applications. The unit features an energy-efficient LED light source housed within a corrosion-resistant, NEMA 4X rated enclosure, ensuring protection in harsh environments.
  • Features a long-lasting LED light source with a 148,000 hr median life
  • Configurable for steady burning or flashing operation with a flash rate of 65 fpm
  • Constructed with a shatter-resistant polycarbonate lens and a 33% glass-filled nylon base
  • Rated NEMA 4X for corrosion resistance
  • Designed for conduit or panel mounting with 1/2 in. internal and 3/4 in. external hub threading
  • Immune to shock and vibration
  • UL and cUL listed