Learn More
Grainger Edwards Signaling LED Warning Light, Red, 120V AC, 1 ea
Catalog No. 502791194
Encompass_Preferred
The 105XBRM Series is a heavy-duty, reliable LED visual signal designed for demanding industrial and marine applications. This beacon features a double Fresnel lens constructed from shatter-resistant polycarbonate and a base made of glass-reinforced thermoplastic polyester resin. It provides versatile operation with both steady-on and 65 fpm flashing modes, supported by an enclosure rated for NEMA 4X, making it suitable for hazardous locations.
  • Shatter-resistant, shock-proof, and vibration-proof design
  • Multi-mode operation with steady-on or 65 fpm flashing
  • UL listed Type 4X and marine-rated enclosure
  • Projected LED lifespan of 148,000 hours
  • Operating temperature range of -35°C to 100°C
  • Suitable for Class I, Division 2, Groups A, B, C, D; Class II, Division 2, Groups F, G; and Class III hazardous locations
Catalog No. 50-279-1194 Supplier Grainger Supplier No. 16G641
The 105XBRM Series is a heavy-duty, reliable LED visual signal designed for demanding industrial and marine applications. This beacon features a double Fresnel lens constructed from shatter-resistant polycarbonate and a base made of glass-reinforced thermoplastic polyester resin. It provides versatile operation with both steady-on and 65 fpm flashing modes, supported by an enclosure rated for NEMA 4X, making it suitable for hazardous locations.
  • Shatter-resistant, shock-proof, and vibration-proof design
  • Multi-mode operation with steady-on or 65 fpm flashing
  • UL listed Type 4X and marine-rated enclosure
  • Projected LED lifespan of 148,000 hours
  • Operating temperature range of -35°C to 100°C
  • Suitable for Class I, Division 2, Groups A, B, C, D; Class II, Division 2, Groups F, G; and Class III hazardous locations