Learn More
Grainger Edwards Signaling Warning Light, LED, Clear, 120V AC, 1 ea
Catalog No. 502791458
Encompass_Preferred
This multimode warning light provides high-visibility signaling for industrial monitoring and alarm applications. Designed for direct hardwired control, the unit features steady-on and lightburst operational modes to alert personnel to hazardous conditions. Constructed from durable, glass-filled nylon with a polycarbonate lens, it is built to withstand demanding environments.
  • Chemical, corrosion, heat, and shatter resistant housing
  • Light source life of 148,000 hours
  • NEMA 4X enclosure rating for protection against environmental hazards
  • Integrated 1/2 in. internal and 3/4 in. external NPT conduit mounting
  • Flash rate of 420 FPM
  • Operational temperature range of -35°C to +66°C (-31°F to +150°F)
  • UL and cUL listed
Catalog No. 50-279-1458 Supplier Grainger Supplier No. 16G638
This multimode warning light provides high-visibility signaling for industrial monitoring and alarm applications. Designed for direct hardwired control, the unit features steady-on and lightburst operational modes to alert personnel to hazardous conditions. Constructed from durable, glass-filled nylon with a polycarbonate lens, it is built to withstand demanding environments.
  • Chemical, corrosion, heat, and shatter resistant housing
  • Light source life of 148,000 hours
  • NEMA 4X enclosure rating for protection against environmental hazards
  • Integrated 1/2 in. internal and 3/4 in. external NPT conduit mounting
  • Flash rate of 420 FPM
  • Operational temperature range of -35°C to +66°C (-31°F to +150°F)
  • UL and cUL listed