Learn More
Grainger Edwards Signaling Warning Light, LED, Dual-Mode, 120V AC, 1 ea
Catalog No. 502788918
Encompass_Preferred
The 125 Class XBR XTRA-BRITE LED beacon is a dual mode visual signaling device designed for indoor and outdoor applications. This 120V AC unit features a corrosion-resistant Type 4X enclosure with a 33% glass-filled nylon base for high heat and chemical resistance. It is engineered for versatile installation, supporting both panel and conduit mounting.
  • Dual mode operation offers steady-on or 65 fpm flashing patterns
  • Durable shatter-resistant polycarbonate lens
  • Immune to shock and vibration for demanding environments
  • Long-lasting LED light source with a projected lifespan of 148,000 hours
  • Operating temperature range of -35°C to 66°C (-31°F to 150°F)
  • Includes double-threaded 1/2 in. internal and 3/4 in. external conduit hub
  • UL and cUL listed for safety and performance
Catalog No. 50-278-8918 Supplier Grainger Supplier No. 16G617
The 125 Class XBR XTRA-BRITE LED beacon is a dual mode visual signaling device designed for indoor and outdoor applications. This 120V AC unit features a corrosion-resistant Type 4X enclosure with a 33% glass-filled nylon base for high heat and chemical resistance. It is engineered for versatile installation, supporting both panel and conduit mounting.
  • Dual mode operation offers steady-on or 65 fpm flashing patterns
  • Durable shatter-resistant polycarbonate lens
  • Immune to shock and vibration for demanding environments
  • Long-lasting LED light source with a projected lifespan of 148,000 hours
  • Operating temperature range of -35°C to 66°C (-31°F to 150°F)
  • Includes double-threaded 1/2 in. internal and 3/4 in. external conduit hub
  • UL and cUL listed for safety and performance