Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Checkers Safety™ Warning Whip, 12 ft, Quick Disconnect, 1 ea

Catalog No. 502791456
Encompass_Preferred

This 12 ft. general-purpose warning whip provides enhanced visibility for vehicles in hazardous work environments. It is constructed from premium fiberglass resin for durability and features a fluorescent nylon mesh flag to ensure high visibility. The system utilizes a quick disconnect base for rapid installation and removal, making it a versatile safety solution for industrial applications.

  • Flag size: 12 in. x 11 in.
  • Flag material: heavy duty Day Bright™ fluorescent nylon mesh
  • Whip material: premium fiberglass resin
  • Base type: quick disconnect
  • Includes debris cap for protection when detached
  • Non-lighted design

Catalog No. 50-279-1456 Supplier Grainger Supplier No. 16D780
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This 12 ft. general-purpose warning whip provides enhanced visibility for vehicles in hazardous work environments. It is constructed from premium fiberglass resin for durability and features a fluorescent nylon mesh flag to ensure high visibility. The system utilizes a quick disconnect base for rapid installation and removal, making it a versatile safety solution for industrial applications.

  • Flag size: 12 in. x 11 in.
  • Flag material: heavy duty Day Bright™ fluorescent nylon mesh
  • Whip material: premium fiberglass resin
  • Base type: quick disconnect
  • Includes debris cap for protection when detached
  • Non-lighted design

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.