Learn More
Grainger Checkers Warning Whip, Non-Lighted, Fiberglass Resin, 8 ft, Reflective X Flag, 1 ea
Catalog No. 502791973
Encompass_Preferred
This non-lighted warning whip is designed for high visibility in industrial environments. Constructed from premium fiberglass resin, this 8 ft whip provides durable performance and includes a reflective X flag for enhanced safety. The system features a threaded hex base for secure mounting.
  • Premium fiberglass resin construction
  • 8 ft length
  • Includes reflective X flag
  • Fluorescent nylon mesh flag
  • Threaded hex base for secure installation
  • Not lighted
Catalog No. 50-279-1973 Supplier Grainger Supplier No. 16D809
This non-lighted warning whip is designed for high visibility in industrial environments. Constructed from premium fiberglass resin, this 8 ft whip provides durable performance and includes a reflective X flag for enhanced safety. The system features a threaded hex base for secure mounting.
  • Premium fiberglass resin construction
  • 8 ft length
  • Includes reflective X flag
  • Fluorescent nylon mesh flag
  • Threaded hex base for secure installation
  • Not lighted