Learn More
Grainger Checkers Safety™ Warning Whip, Non-Lighted, Nylon Mesh, 8 ft., 1 ea.
Catalog No. 502792316
Encompass_Preferred
This general-purpose, non-lighted warning whip is designed to enhance safety and visibility at worksites. The 8 ft. assembly features a durable fiberglass whip and a high-visibility orange nylon mesh flag with a reflective white X pattern.
  • Equipped with a quick disconnect base for easy installation and removal
  • Features a heavy-duty fluorescent nylon mesh flag for maximum visibility
  • Includes a debris cap to protect the base connection
  • Constructed from premium fiberglass resin for durability in demanding environments
  • Reflective X pattern provides enhanced safety in low-light conditions
Catalog No. 50-279-2316 Supplier Grainger Supplier No. 16D808
This general-purpose, non-lighted warning whip is designed to enhance safety and visibility at worksites. The 8 ft. assembly features a durable fiberglass whip and a high-visibility orange nylon mesh flag with a reflective white X pattern.
  • Equipped with a quick disconnect base for easy installation and removal
  • Features a heavy-duty fluorescent nylon mesh flag for maximum visibility
  • Includes a debris cap to protect the base connection
  • Constructed from premium fiberglass resin for durability in demanding environments
  • Reflective X pattern provides enhanced safety in low-light conditions