Learn More
Diagnostic Biosystems WARTHIN-STARRY Warthin-Starry
Catalog No. 502407435
Encompass_Preferred
The Warthin-Starry Stain Kit is intended for use in the visualization of Spirochetes, Helicobacter pylori, Legionella pneumophila, and Cat Scratch Fever bacteria. This kit may be used on formalin-fixed, paraffin-embedded sections.
Catalog No. 50-240-7435 Supplier Diagnostic Biosystems Supplier No. KT036
The Warthin-Starry Stain Kit is intended for use in the visualization of Spirochetes, Helicobacter pylori, Legionella pneumophila, and Cat Scratch Fever bacteria. This kit may be used on formalin-fixed, paraffin-embedded sections.