Learn More
Bachem Fmoc-Cys(StBu)-OH
SDP
Supplier:  Bachem 4007445.0100
Encompass_Preferred
Fmoc-Cys(StBu)-OH, 73724-43-3, C22H25NO4S2, Mr 431.58, 100g. Was converted into the corresponding N-Me-Cys derivative in excellent yield. Educt for obtaining the Cys 4-methyl-2,6,7-trioxabicyclo[2,2,2]octyl (OBO) orthoester. The OBO ester was attached to a thiol resin as disulfide for obtaining peptides containing a C-terminal cysteine.
Catalog No. 50-194-7139