Learn More
Aviva Systems Biology Corp Wasf2 Antibody - N-terminal region (ARP51323_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51323_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Wasf2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AW742646, D4Ertd13e, WAVE2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-576