Learn More
Aviva Systems Biology Corp WASF3 Antibody - N-terminal region (ARP52219_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52219_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WASF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WASF3. Alternate names: Brush-1, KIAA0900, SCAR3, WAVE3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60704