Learn More
Perkin Elmer US LLC Wash and Waste Vial Diffuser, conical shape
Catalog No. 502104261
Encompass_Preferred
The conical shape of the PerkinElmer diffusers allow the needle to penetrate into solvent or waste. Reduces spillage and large losses through evaporation.
Catalog No. 50-210-4261 Supplier Perkin Elmer US LLC Supplier No. N6101276
The conical shape of the PerkinElmer diffusers allow the needle to penetrate into solvent or waste. Reduces spillage and large losses through evaporation.