Learn More
Electron Microscopy Sciences GHS Multi-lingual Wash Bottle Acetone, 250 ml, 145 mm height, 60 mm diameter
SDP
Catalog No. 501928524
Encompass_Preferred
Multi-lingual wide mouth wash bottles featuring the patented DripLok vapor venting valve that prevents solvent drips. Indelibly printed in English, French, German and Spanish. Choose from four solvent labels, all including the new Globally Harmonized Symbols, NFPA diamond, risk phrases and safety information. Fitted with color coded one-piece PP closures that maintain their shape in use. Closures feature precision jet-tip aperture for fine control.
Volume: 250 ml
Height: 145 mm
Diameter: 60 mm
Catalog No. 50-192-8524 Supplier Electron Microscopy Sciences Supplier No. 64100
Multi-lingual wide mouth wash bottles featuring the patented DripLok vapor venting valve that prevents solvent drips. Indelibly printed in English, French, German and Spanish. Choose from four solvent labels, all including the new Globally Harmonized Symbols, NFPA diamond, risk phrases and safety information. Fitted with color coded one-piece PP closures that maintain their shape in use. Closures feature precision jet-tip aperture for fine control.
Volume: 250 ml
Height: 145 mm
Diameter: 60 mm