Learn More
Chemglass Life Sciences Bottle, Washing, Safety, Polyethylene, 500mL, Toluene
SDP
Catalog No. CG113014
Encompass_Preferred
Bottle, Washing, Safety, Polyethylene, 500mL, Toluene. Easy-to-read, low-density polyethylene dispensing bottles are break-resistantIntended for short-term dispensing useInclude DOT, CAS numbers, NFPA codes and symbols, protective equipment symbols, and target organ informationEasy-to-read, break-resistant dispensing bottles include DOT, CAS numbers, National Fire Protection Association (NFPA) codes and symbols, protective equipment symbols, and target organ information. Polypropylene nozzle assemblies are leakproof and color-coded to match bottle label. Toluene bottle is solid red with white lettering and hazard information.
Catalog No. CG113014 Supplier Chemglass Life Sciences Supplier No. CG113014
Bottle, Washing, Safety, Polyethylene, 500mL, Toluene. Easy-to-read, low-density polyethylene dispensing bottles are break-resistantIntended for short-term dispensing useInclude DOT, CAS numbers, NFPA codes and symbols, protective equipment symbols, and target organ informationEasy-to-read, break-resistant dispensing bottles include DOT, CAS numbers, National Fire Protection Association (NFPA) codes and symbols, protective equipment symbols, and target organ information. Polypropylene nozzle assemblies are leakproof and color-coded to match bottle label. Toluene bottle is solid red with white lettering and hazard information.