Learn More
Grainger 125 mL Wash Bottles
Catalog No. NC2194437
Encompass_Preferred
The Grainger 125 mL Polypropylene Wash Bottle is made of durable polypropylene plastic with a leak-proof screw cap. It has a tapered tip for controlled dispensing and is ideal for dispensing solvents and reagents. The bottle has a capacity of 125 mL and is autoclavable for sterilization. It can withstand temperatures up to 121°C. The dimensions are 3" diameter x 8-1/2" height. This wash bottle provides reliable dispensing for various laboratory applications.
This item is not returnable. View return policy
Catalog No. NC2194437 Supplier Grainger Supplier No. 6FAV3
The Grainger 125 mL Polypropylene Wash Bottle is made of durable polypropylene plastic with a leak-proof screw cap. It has a tapered tip for controlled dispensing and is ideal for dispensing solvents and reagents. The bottle has a capacity of 125 mL and is autoclavable for sterilization. It can withstand temperatures up to 121°C. The dimensions are 3" diameter x 8-1/2" height. This wash bottle provides reliable dispensing for various laboratory applications.