Learn More
Chemglass Life Sciences Bottle, Washing, Safety, Polyethylene, 500mL, Methanol
SDP
Catalog No. CG113011
Encompass_Preferred
Bottle, Washing, Safety, Polyethylene, 500mL, Methanol. Easy-to-read, low-density polyethylene dispensing bottles are break-resistantIntended for short-term dispensing useInclude DOT, CAS numbers, NFPA codes and symbols, protective equipment symbols, and target organ informationEasy-to-read, break-resistant dispensing bottles include DOT, CAS numbers, National Fire Protection Association (NFPA) codes and symbols, protective equipment symbols, and target organ information. Polypropylene nozzle assemblies are leakproof and color-coded to match bottle label. Toluene bottle is solid red with white lettering and hazard information.
Catalog No. CG113011 Supplier Chemglass Life Sciences Supplier No. CG113011
Bottle, Washing, Safety, Polyethylene, 500mL, Methanol. Easy-to-read, low-density polyethylene dispensing bottles are break-resistantIntended for short-term dispensing useInclude DOT, CAS numbers, NFPA codes and symbols, protective equipment symbols, and target organ informationEasy-to-read, break-resistant dispensing bottles include DOT, CAS numbers, National Fire Protection Association (NFPA) codes and symbols, protective equipment symbols, and target organ information. Polypropylene nozzle assemblies are leakproof and color-coded to match bottle label. Toluene bottle is solid red with white lettering and hazard information.