Learn More
Chemglass Life Sciences Bottle, Washing, Safety, Polyethylene, 500mL, Isopropanol
SDP
Catalog No. CG113008
Encompass_Preferred
Bottle, Washing, Safety, Polyethylene, 500mL, Isopropanol. Easy-to-read, low-density polyethylene dispensing bottles are break-resistantIntended for short-term dispensing useInclude DOT, CAS numbers, NFPA codes and symbols, protective equipment symbols, and target organ informationEasy-to-read, break-resistant dispensing bottles include DOT, CAS numbers, National Fire Protection Association (NFPA) codes and symbols, protective equipment symbols, and target organ information. Polypropylene nozzle assemblies are leakproof and color-coded to match bottle label. Toluene bottle is solid red with white lettering and hazard information.
Catalog No. CG113008 Supplier Chemglass Life Sciences Supplier No. CG113008
Bottle, Washing, Safety, Polyethylene, 500mL, Isopropanol. Easy-to-read, low-density polyethylene dispensing bottles are break-resistantIntended for short-term dispensing useInclude DOT, CAS numbers, NFPA codes and symbols, protective equipment symbols, and target organ informationEasy-to-read, break-resistant dispensing bottles include DOT, CAS numbers, National Fire Protection Association (NFPA) codes and symbols, protective equipment symbols, and target organ information. Polypropylene nozzle assemblies are leakproof and color-coded to match bottle label. Toluene bottle is solid red with white lettering and hazard information.