Learn More
Enzo Life Sciences Wash buffer concentrate, (100ml)
SDP
Catalog No. 502006815
Encompass_Preferred
Formulation: Liquid. TRIS buffered saline containing detergents. Long Term Storage: Ambient. Applications: ELISA .
Catalog No. 50-200-6815 Supplier Enzo Life Sciences Supplier No. ADI801287
Formulation: Liquid. TRIS buffered saline containing detergents. Long Term Storage: Ambient. Applications: ELISA .