Learn More
Abcam 10X Wash Buffer PT, 1000 mL, concentrated format, ELISA wash buffer, dilute to 1X with deionized water
Supplier:  Abcam AB2069771000ML
Encompass_Preferred
Abcam's 10X Wash Buffer PT is a concentrated ELISA wash buffer designed to enhance performance with SimpleStep ELISA kits and Matched Antibody Pair kits. Its ability to be diluted to a 1X solution using deionized water not only provides flexibility but also promotes efficient cost management. With a reliable and optimized composition, this product is integral for achieving consistent assay results in various immunoassay systems.
  • Concentrated format enables dilution to 1X
  • Composed for reliable washing performance
  • Compatible with SimpleStep ELISA and Matched Antibody Pair kits
  • Recommended dilution with deionized water
  • Shipping condition on Blue Ice
  • Short and long-term storage at +4°C
  • For research use only
Catalog No. NC1564749