Learn More
Enzo Life Sciences Wash buffer salts (3 packs) (3L)
SDP
Catalog No. 502012501
Encompass_Preferred
Wash Buffer Salts can be used in tissue preparations, immunohistochemical staining procedures and in assays that require HRP-linked detection reagents. Each packet of salts yields 1 liter of buffered saline. Formulation: Lyophilized. Long Term Storage: Ambient.
Catalog No. 50-201-2501 Supplier Enzo Life Sciences Supplier No. ENZ33802
Wash Buffer Salts can be used in tissue preparations, immunohistochemical staining procedures and in assays that require HRP-linked detection reagents. Each packet of salts yields 1 liter of buffered saline. Formulation: Lyophilized. Long Term Storage: Ambient.