Learn More
Research Products International Corp Wash-N-Dry Coverslip RK
SDP
Catalog No. 502229828
Encompass
Wash-N-Dry Coverslip Rack offers fast and easy cleaning, surface-treating and drying of microscope cover slips. Rack holds up to ten 18, 22, and 25mm coverslips. Manufactured from one piece glass infused polypropylene for durability. Ideal rack fits inside 100 ml beaker and allows space for small magnetic stirring bar.
Convenient
Holds 10 coverslips
Fits inside 100 ml beaker
Catalog No. 50-222-9828 Supplier Research Products International Corp Supplier No. 249410
Wash-N-Dry Coverslip Rack offers fast and easy cleaning, surface-treating and drying of microscope cover slips. Rack holds up to ten 18, 22, and 25mm coverslips. Manufactured from one piece glass infused polypropylene for durability. Ideal rack fits inside 100 ml beaker and allows space for small magnetic stirring bar.
Convenient
Holds 10 coverslips
Fits inside 100 ml beaker