Learn More
Sigma Aldrich Fine Chemicals Biosciences Wash-N-DryTM coverslip rac1EA
Catalog No. 502900253
Encompass
Holds 10 Coverslips fast and easy cleaning surface-treating and drying. Holds all slipcover sizes including 18 22 and 25 mm.Durable and Hassle free One piece polypropylene rack does not require assembly and is chemically and thermally resistant. Autoclavable.Secure Innovative design includes a notch to secure coverslips with a rubber band or loop of mono-filament.Compatible fits inside a 100 mL beaker with space underneath for a small stir bar when submerged.Note The gap at the bottom of the Wash-N-Dry rack measures 8 mm (round coverslips also work with this product).
Catalog No. 50-290-0253 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6885681EA
Holds 10 Coverslips fast and easy cleaning surface-treating and drying. Holds all slipcover sizes including 18 22 and 25 mm.Durable and Hassle free One piece polypropylene rack does not require assembly and is chemically and thermally resistant. Autoclavable.Secure Innovative design includes a notch to secure coverslips with a rubber band or loop of mono-filament.Compatible fits inside a 100 mL beaker with space underneath for a small stir bar when submerged.Note The gap at the bottom of the Wash-N-Dry rack measures 8 mm (round coverslips also work with this product).