Learn More
Electron Microscopy Sciences Wash-Off™ Pre-Printed Label Warning: Poison, 25 x 25 mm
SDP
Catalog No. 5033387
Encompass_Preferred
Pre-printed Wash-Off Labels make it easy to mark bottles containing harmful chemicals or reagents. These convenient labels will save time and improve the safety of every lab. Wash the labels away when the label is no longer needed.
Size: 25 x 25 mm
Catalog No. 50-333-87 Supplier Electron Microscopy Sciences Supplier No. 7182622
Pre-printed Wash-Off Labels make it easy to mark bottles containing harmful chemicals or reagents. These convenient labels will save time and improve the safety of every lab. Wash the labels away when the label is no longer needed.
Size: 25 x 25 mm