Learn More
Siemens Healthineers Wash Solution
Catalog No. 23044708 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 15 mL
Catalog No. Quantity
23-044-708 10 x 15 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-708 Supplier Siemens Healthineers Supplier No. 10446684
Add to Cart
Add to Cart
This item is not returnable. View return policy
Wash Solution is used for cleaning the Siemens Healthcare Diagnostics Coagulation Analyzers.
TRUSTED_SUSTAINABILITY
Specifications
Type Wash Solution
For Use With (Equipment) Coagulation Analyzers
Quantity 10 x 15 mL