Learn More
KPM ANALYTICS NORTH AMERICA CORPORATION KPM Analytics Cleaning Solution, Removes Residues, Concentrated Solution, 4x133 mL Bottles
Catalog No. NC3002242
Encompass
KPM Analytics Cleaning Solution is an effective cleaning agent designed for the removal of liquid residues from laboratory instruments. Supplied as a concentrated 4-pack of 133 mL bottles, it ensures optimal optical purity and performance of analyzers like the SmartChem 200. Its composition, including less than 1% sodium hydroxide, minimizes interference in analysis while promoting thorough cleaning.
Catalog No. NC3002242 Supplier KPM ANALYTICS NORTH AMERICA CORPORATION Supplier No. 650524400
KPM Analytics Cleaning Solution is an effective cleaning agent designed for the removal of liquid residues from laboratory instruments. Supplied as a concentrated 4-pack of 133 mL bottles, it ensures optimal optical purity and performance of analyzers like the SmartChem 200. Its composition, including less than 1% sodium hydroxide, minimizes interference in analysis while promoting thorough cleaning.