Learn More
Seracare Life Sciences Inc WASH SOLUTION CONCENTRATE KIT
Catalog No. 5067410
Encompass_Preferred
Convenient 20X concentrate containing imidazole-buffered saline and Tween 20. Kit contains 4 x 200 ml bottles, enough to prepare 16 liters of working solution. Suitable for ELISA, blotting and immunohistochemistry applications.
This item is not returnable. View return policy
Catalog No. 50-674-10 Supplier Seracare Life Sciences Inc Supplier No. 51500008
Convenient 20X concentrate containing imidazole-buffered saline and Tween 20. Kit contains 4 x 200 ml bottles, enough to prepare 16 liters of working solution. Suitable for ELISA, blotting and immunohistochemistry applications.