Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wash Solution, For Surfactants, LabChem™

Catalog No. LC267002 Shop All LabChem Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC267002 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC267002 Supplier LabChem, Inc. Supplier No. LC267002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Wash Solution
Color Colorless
Name Note For Surfactants
CAS Max % 94.97, 4.35, 0.68
CAS 7558-80-7, 7664-93-9
Linear Formula NaH2PO4
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 119.98
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Phosphorus oxides; Sulfur compounds
Quantity 1 L
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.