Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Wash Solution for GeneJET Plasmid Miniprep Kit

Catalog No. R1243
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R1243 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R1243 Supplier Thermo Scientific™ Supplier No. R1243
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

Thermo Scientific GeneJET Wash Solution is a component of the GeneJET Plasmid Miniprep Kit and may be purchased separately. Provided in 100 mL aliquots.

Thermo Scientific GeneJET Wash Solution is a component of the GeneJET Plasmid Miniprep Kit (Cat. No. K0502/K0503) and may be purchased separately. Provided in 100 mL aliquots.

Thermo Scientific GeneJET Wash Solution is a component of the GeneJET Plasmid Miniprep Kit (Cat. No. K0502 and K0503) and may be purchased separately. Provided in 100 mL aliquots.

Related Products

  • Binding Buffer for GeneJET Plasmid Miniprep Kit (Cat. No. R1331)
  • GeneJET NGS Cleanup Kit (K0851)
  • Binding Buffer for GeneJET NGS Cleanup Kit (R2811)
  • Prewash Buffer for GeneJET NGS Cleanup Kit (R2821)
  • Wash Buffer for GeneJET NGS Cleanup Kit (R2831)
  • GeneJET Plasmid Miniprep Kit (K0502)
  • RNase A (R1253)
  • Elution Buffer for GeneJET Plasmid Miniprep Kit (R1263)
  • Lysis Solution for GeneJET Plasmid Miniprep Kit (R1223)
  • Resuspension Solution for GeneJET Plasmid Miniprep Kit (R1213)
  • Neutralization Solution for GeneJET Plasmid Miniprep Kit (R1233)
TRUSTED_SUSTAINABILITY

Specifications

Buffer Wash Buffer
Format Kit
Product Line GeneJET
Product Type Wash Solution
Quantity 100 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.