Learn More
CELLULAR TECHNOLOGY LTD CTL-Wash™ Supplement 10x, serum-free formulation
SDP
Catalog No. nc9711918
Encompass
Ctl-Wash Is A Formulation Specially Made To Keep Pbmc In A Nutrient-Rich Environment While Maintaining Their Full Viability And Functionality During Washing. The Two To Three Washing Steps Required After The Ficoll Isolation Or During Thawing Of Pbmc And The Time Required For Their Counting Typically Adds Up To 30-40 Minutes. During This Time, The Pbmc Quality Will Deteriorate If Kept In A Nutritionally-Deficient Environment. If Uncharacterized Serum Is Added, Mitogenic Or Toxic Effects Can Affect The Cells. Ctl-Wash Supplement Is A 10X Serum-Free Formulation Which Has To Be Added To Rpmi-1640. It Contains Injection-Grade Water Supplemented With Amino Acids, Vitamins, Inorganic Salts, And Human Serum Albumin (Hsa) In A Proprietary Composition To Provide A Balanced Environment For Maximizing Pbmc Survival. Ctl-Wash Supplement 10X Is Sold In Convenient Sterile 100Ml Units Ready For Dilution.
Catalog No. NC9711918 Supplier CELLULAR TECHNOLOGY LTD Supplier No. CTLW010
Ctl-Wash Is A Formulation Specially Made To Keep Pbmc In A Nutrient-Rich Environment While Maintaining Their Full Viability And Functionality During Washing. The Two To Three Washing Steps Required After The Ficoll Isolation Or During Thawing Of Pbmc And The Time Required For Their Counting Typically Adds Up To 30-40 Minutes. During This Time, The Pbmc Quality Will Deteriorate If Kept In A Nutritionally-Deficient Environment. If Uncharacterized Serum Is Added, Mitogenic Or Toxic Effects Can Affect The Cells. Ctl-Wash Supplement Is A 10X Serum-Free Formulation Which Has To Be Added To Rpmi-1640. It Contains Injection-Grade Water Supplemented With Amino Acids, Vitamins, Inorganic Salts, And Human Serum Albumin (Hsa) In A Proprietary Composition To Provide A Balanced Environment For Maximizing Pbmc Survival. Ctl-Wash Supplement 10X Is Sold In Convenient Sterile 100Ml Units Ready For Dilution.