Learn More
Waters Corp Waters Corporation Wash Tube Seal, 4 per Pack, for HPLC Systems
Catalog No. 502039978 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These wash tube seals are an integral component of the solvent management system found in 2695 Separations Modules. They are part of the seal wash system, designed to maximize the lifespan of plunger seals.
  • Integral to the solvent management system of 2695 Separations Modules.
  • Maximizes plunger seal life.
  • Kit contains four seals.
  • Covers both pump heads.
Catalog No. 50-203-9978 Supplier Waters Corp Supplier No. WAT270940
These wash tube seals are an integral component of the solvent management system found in 2695 Separations Modules. They are part of the seal wash system, designed to maximize the lifespan of plunger seals.
  • Integral to the solvent management system of 2695 Separations Modules.
  • Maximizes plunger seal life.
  • Kit contains four seals.
  • Covers both pump heads.