Learn More
Beckman Coulter Beckman Coulter Wash WBD reagent (C39489)
Catalog No. NC2164210
Encompass
Beckman Coulter Wash WBD (C39489) is a liquid wash reagent used with Beckman Coulter diagnostic analyzers in automated wash cycles to remove residual reagents and help maintain assay performance.
  • Provides instrument wash for automated analyzers.
  • Helps remove residual reagent and reduce carryover.
  • Intended for laboratory diagnostic use.
  • Supplied as a liquid reagent for use in instrument wash systems.
This item is not returnable. View return policy
Catalog No. NC2164210 Supplier Beckman Coulter Supplier No. C39489
Beckman Coulter Wash WBD (C39489) is a liquid wash reagent used with Beckman Coulter diagnostic analyzers in automated wash cycles to remove residual reagents and help maintain assay performance.
  • Provides instrument wash for automated analyzers.
  • Helps remove residual reagent and reduce carryover.
  • Intended for laboratory diagnostic use.
  • Supplied as a liquid reagent for use in instrument wash systems.