Learn More
LAMPIRE Biological Laboratories Guinea Pig Washed Red Blood Cells, 5% Concentration, QC Tested, 50 mL
SDP
Catalog No. NC3443360
Encompass
Washed Guinea Pig Red Blood Cells are aseptically collected and processed to ensure a 5% concentration in Alsever's solution. This product is ideal for in vitro applications in hematology and immunology, providing consistent and reliable quality upon order. It is designed for medical research and diagnostics, making it suitable for various assay techniques while ensuring viability for experiments.
Catalog No. NC3443360 Supplier LAMPIRE Biological Laboratories Supplier No. 7243108
Washed Guinea Pig Red Blood Cells are aseptically collected and processed to ensure a 5% concentration in Alsever's solution. This product is ideal for in vitro applications in hematology and immunology, providing consistent and reliable quality upon order. It is designed for medical research and diagnostics, making it suitable for various assay techniques while ensuring viability for experiments.