Learn More
Perkin Elmer US LLC Injector Support Adapter Washer
Catalog No. NC1545097
Encompass_Preferred
This washer is designed for use in injector support adapters in PerkinElmer ICP-OES instruments.
Catalog No. NC1545097 Supplier Perkin Elmer US LLC Supplier No. N0776014
This washer is designed for use in injector support adapters in PerkinElmer ICP-OES instruments.