Learn More
Microsolv Technology Corporation Washers, for use with Low Pressure, Tubing Connectors. Use with Flanged, 1/16th", low pressure tubing. Polypropylene. 10/PK.
Catalog No. 502271415
Encompass_Preferred
Use with "Male Nuts" for low pressure tubing connections in HPLC or other fluidic connections.
This item is not returnable. View return policy
Catalog No. 50-227-1415 Supplier Microsolv Technology Corporation Supplier No. 45016WASHER
Use with "Male Nuts" for low pressure tubing connections in HPLC or other fluidic connections.