Learn More
Microsolv Technology Corporation Washers, for use with Low Pressure, Tubing Connectors. Use with Flanged, 1/8th", tubing. Polypropylene. 10/PK.
Supplier:  Microsolv Technology Corporation 45008WASHER
Encompass_Preferred
These washers are made of high quality plastic (polypropylene) for use with Male Nuts (connectors) for low pressure 1/8th inch tubing. Washers are required with Flanged Tubing Connectors.
Catalog No. 50-227-1409