Learn More
Analytical Sales & Services Washers
Catalog No. 5000805796
Encompass
Cadmium-Plated Steel Mil. Spec. Washers\nUse to affix a Thermal Adapter Plate (TAP) to a Thermal Transfer Deck (TTD).\nUse with Stainless Steel Screws for TAP (TAPSCREW). Use two washers for each screw.\n\nQty/Pkg: 200
Catalog No. 50-008-05796 Supplier Analytical Sales & Services Supplier No. TAPWASH
Cadmium-Plated Steel Mil. Spec. Washers\nUse to affix a Thermal Adapter Plate (TAP) to a Thermal Transfer Deck (TTD).\nUse with Stainless Steel Screws for TAP (TAPSCREW). Use two washers for each screw.\n\nQty/Pkg: 200