Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arkray Washing Solution for Tubes
SDP

Catalog No. 21377284
Change view
Click to view available options
Sample Type:
Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
21-377-284 Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For laboratory use only. Avoid contact with eyes and skin. Use in a well-ventilated area and wear appropriate protective equipment.
Catalog No. 21-377-284 Supplier Arkray Supplier No. 71237
Add to Cart
Add to Cart
For laboratory use only. Avoid contact with eyes and skin. Use in a well-ventilated area and wear appropriate protective equipment.

Washing Solution for Tubes helps keep ARKRAY analyzers performing at their best by thoroughly cleaning sample tubes to prevent carryover and maintain accurate results. This reliable wash reagent supports consistent system performance.

  • Removes sample residue to maintain clean tubing pathways.
  • Reduces carryover and supports accurate analyzer results.
  • Prevents clogs and helps minimize instrument downtime.
  • Ready-to-use formula designed for ARKRAY analyzer systems.
  • Supports consistent performance in routine maintenance cycles.

Specifications

Sample Type Urine
Type Solution
Form Liquid

Compatible with ARKRAY analyzer maintenance protocols. For optimal performance, use as part of the recommended cleaning schedule.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.