Learn More
Grainger Washroom Mirror 18 in W 36 in H
Catalog No. 502753507
Encompass_Preferred
The Ketcham Washroom Mirror Rectangular is a rectangular bathroom mirror that measures 24 inches wide by 36 inches tall. It has a 1 inch beveled edge and is made from distortion-free plate glass with a silver vacuum coating. The mirror can be hung vertically or horizontally using the pre-drilled holes and mounting hardware. It is designed for commercial restrooms, schools, hotels, and other high-traffic areas. The silver coating provides over 90% light reflectivity for a bright, clear reflection. This durable mirror resists corrosion and features a 5 year warranty.
Catalog No. 50-275-3507 Supplier Grainger Supplier No. 14C574
The Ketcham Washroom Mirror Rectangular is a rectangular bathroom mirror that measures 24 inches wide by 36 inches tall. It has a 1 inch beveled edge and is made from distortion-free plate glass with a silver vacuum coating. The mirror can be hung vertically or horizontally using the pre-drilled holes and mounting hardware. It is designed for commercial restrooms, schools, hotels, and other high-traffic areas. The silver coating provides over 90% light reflectivity for a bright, clear reflection. This durable mirror resists corrosion and features a 5 year warranty.